Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612413 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf71 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C11orf71 - chromosome 11 open reading frame 71 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBM7 - RNA binding motif protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286045.1 | Intron | NP_001272974.1 | ||||
NM_001286046.1 | Intron | NP_001272975.1 | ||||
NM_001286047.1 | Intron | NP_001272976.1 | ||||
NM_001286048.1 | Intron | NP_001272977.1 | ||||
NM_016090.3 | Intron | NP_057174.1 |
Set Membership: |
HapMap |