Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGTGGTTCTCCCATACAACTGA[C/T]GTGTTTTTCACCTCAAGAAACCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612135 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CABYR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CABYR - calcium binding tyrosine phosphorylation regulated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308231.1 | Intron | NP_001295160.1 | ||||
NM_012189.3 | Intron | NP_036321.2 | ||||
NM_138643.2 | Intron | NP_619584.1 | ||||
NM_138644.2 | Intron | NP_619585.1 | ||||
NM_153768.2 | Intron | NP_722452.1 | ||||
NM_153769.2 | Intron | NP_722453.1 | ||||
NM_153770.2 | Intron | NP_722454.1 | ||||
XM_005258247.2 | Intron | XP_005258304.1 | ||||
XM_017025706.1 | Intron | XP_016881195.1 |
TTC39C - tetratricopeptide repeat domain 39C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |