Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACACAGCTGCCTTGCCTTCACT[C/T]GATTATCCTGTCTGAAGTTTAACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC39C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTC39C - tetratricopeptide repeat domain 39C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135993.1 | Intron | NP_001129465.1 | ||||
NM_001243425.1 | Intron | NP_001230354.1 | ||||
NM_001292030.1 | Intron | NP_001278959.1 | ||||
NM_153211.3 | Intron | NP_694943.2 | ||||
XM_005258198.4 | Intron | XP_005258255.1 | ||||
XM_006722392.3 | Intron | XP_006722455.1 | ||||
XM_011525814.2 | Intron | XP_011524116.1 | ||||
XM_017025550.1 | Intron | XP_016881039.1 |
TTC39C-AS1 - TTC39C antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |