Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGGGCAGGGGAGGGAGTGGCGA[A/G]CGGCTGCCGGAGAGGCGGGAAGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603265 MIM: 604161 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARID3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARID3A - AT-rich interaction domain 3A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KISS1R - KISS1 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032551.4 | 562 | Intron | NP_115940.2 | |||
XM_017027382.1 | 562 | UTR 5 | XP_016882871.1 |
R3HDM4 - R3H domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |