Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGATTCTCTTCTCCCTTCCCGCC[C/T]CCAGGCCCTTGTAGACTCTGGCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610078 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MORC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MORC3 - MORC family CW-type zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320445.1 | Intron | NP_001307374.1 | ||||
NM_001320446.1 | Intron | NP_001307375.1 | ||||
NM_015358.2 | Intron | NP_056173.1 | ||||
XM_011529518.2 | Intron | XP_011527820.1 | ||||
XM_017028311.1 | Intron | XP_016883800.1 | ||||
XM_017028312.1 | Intron | XP_016883801.1 |