Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGGAAAGCATGAGTGGGCTAGG[C/G]GGAGAAGAAGAGCGAAAGAGAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPA5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPA5 - carboxypeptidase A5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127441.1 | Intron | NP_001120913.1 | ||||
NM_001127442.1 | Intron | NP_001120914.1 | ||||
NM_001318223.1 | Intron | NP_001305152.1 | ||||
NM_080385.4 | Intron | NP_525124.3 | ||||
XM_005250710.1 | Intron | XP_005250767.1 | ||||
XM_005250712.1 | Intron | XP_005250769.1 | ||||
XM_011516698.1 | Intron | XP_011515000.1 | ||||
XM_011516701.1 | Intron | XP_011515003.1 | ||||
XM_011516702.1 | Intron | XP_011515004.1 | ||||
XM_011516703.1 | Intron | XP_011515005.1 | ||||
XM_011516704.1 | Intron | XP_011515006.1 | ||||
XM_011516705.1 | Intron | XP_011515007.1 |
LOC101928632 - uncharacterized LOC101928632 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |