Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602622 MIM: 603022 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNASE1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DNASE1L2 - deoxyribonuclease I-like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
E4F1 - E4F transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288776.1 | 490 | Silent Mutation | GCA,GCG | A,A 98 | NP_001275705.1 | |
NM_001288778.1 | 490 | Silent Mutation | GCA,GCG | A,A 98 | NP_001275707.1 | |
NM_004424.4 | 490 | Silent Mutation | GCA,GCG | A,A 98 | NP_004415.3 | |
XM_005255155.1 | 490 | Silent Mutation | GCA,GCG | A,A 151 | XP_005255212.1 | |
XM_005255156.1 | 490 | Silent Mutation | GCA,GCG | A,A 151 | XP_005255213.1 | |
XM_006720858.1 | 490 | Silent Mutation | GCA,GCG | A,A 98 | XP_006720921.1 | |
XM_011522402.1 | 490 | Silent Mutation | GCA,GCG | A,A 151 | XP_011520704.1 | |
XM_011522403.1 | 490 | Silent Mutation | GCA,GCG | A,A 138 | XP_011520705.1 | |
XM_017023010.1 | 490 | Silent Mutation | GCA,GCG | A,A 151 | XP_016878499.1 | |
XM_017023011.1 | 490 | Silent Mutation | GCA,GCG | A,A 138 | XP_016878500.1 |
Set Membership: |
Validated |