Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602313 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OVOL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
OVOL1 - ovo like transcriptional repressor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004561.3 | 776 | Intron | NP_004552.2 | |||
XM_005274018.3 | 776 | Intron | XP_005274075.1 | |||
XM_011545067.2 | 776 | Intron | XP_011543369.1 | |||
XM_011545068.2 | 776 | UTR 5 | XP_011543370.1 | |||
XM_017017837.1 | 776 | Intron | XP_016873326.1 | |||
XM_017017838.1 | 776 | Intron | XP_016873327.1 |
OVOL1-AS1 - OVOL1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |