Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 130620 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPS14 - ribosomal protein S14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025070.1 | 1447 | Intron | NP_001020241.1 | |||
NM_001025071.1 | 1447 | Intron | NP_001020242.1 | |||
NM_005617.3 | 1447 | Intron | NP_005608.1 | |||
XM_017009684.1 | 1447 | UTR 3 | XP_016865173.1 |
Set Membership: |
Validated |