Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600662 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEF2C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MEF2C - myocyte enhancer factor 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001131005.2 | Intron | NP_001124477.1 | ||||
NM_001193347.1 | Intron | NP_001180276.1 | ||||
NM_001193348.1 | Intron | NP_001180277.1 | ||||
NM_001193349.1 | Intron | NP_001180278.1 | ||||
NM_001193350.1 | Intron | NP_001180279.1 | ||||
NM_001308002.1 | Intron | NP_001294931.1 | ||||
NM_002397.4 | Intron | NP_002388.2 | ||||
XM_005248511.2 | Intron | XP_005248568.1 | ||||
XM_006714619.2 | Intron | XP_006714682.1 | ||||
XM_006714625.3 | Intron | XP_006714688.1 | ||||
XM_011543396.2 | Intron | XP_011541698.1 | ||||
XM_011543397.2 | Intron | XP_011541699.1 | ||||
XM_011543400.1 | Intron | XP_011541702.1 | ||||
XM_011543401.1 | Intron | XP_011541703.1 | ||||
XM_017009475.1 | Intron | XP_016864964.1 | ||||
XM_017009476.1 | Intron | XP_016864965.1 | ||||
XM_017009477.1 | Intron | XP_016864966.1 | ||||
XM_017009478.1 | Intron | XP_016864967.1 | ||||
XM_017009479.1 | Intron | XP_016864968.1 | ||||
XM_017009480.1 | Intron | XP_016864969.1 | ||||
XM_017009481.1 | Intron | XP_016864970.1 | ||||
XM_017009482.1 | Intron | XP_016864971.1 | ||||
XM_017009483.1 | Intron | XP_016864972.1 |