Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612687 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RGMB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RGMB - repulsive guidance molecule family member b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012761.2 | Intron | NP_001012779.2 | ||||
XM_005271966.3 | Intron | XP_005272023.1 | ||||
XM_011543345.2 | Intron | XP_011541647.1 | ||||
XM_011543346.2 | Intron | XP_011541648.1 | ||||
XM_011543347.2 | Intron | XP_011541649.2 | ||||
XM_017009390.1 | Intron | XP_016864879.1 | ||||
XM_017009391.1 | Intron | XP_016864880.1 |
RGMB-AS1 - RGMB antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |