Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611304 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM159 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM159 - transmembrane protein 159 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301769.1 | Intron | NP_001288698.1 | ||||
NM_001301771.1 | Intron | NP_001288700.1 | ||||
NM_001301773.1 | Intron | NP_001288702.1 | ||||
NM_001301774.1 | Intron | NP_001288703.1 | ||||
NM_001301775.1 | Intron | NP_001288704.1 | ||||
NM_020422.5 | Intron | NP_065155.3 | ||||
XM_005255439.4 | Intron | XP_005255496.1 | ||||
XM_006721066.2 | Intron | XP_006721129.1 | ||||
XM_006721067.3 | Intron | XP_006721130.1 | ||||
XM_006721069.3 | Intron | XP_006721132.1 | ||||
XM_017023471.1 | Intron | XP_016878960.1 | ||||
XM_017023472.1 | Intron | XP_016878961.1 | ||||
XM_017023473.1 | Intron | XP_016878962.1 | ||||
XM_017023474.1 | Intron | XP_016878963.1 |