Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601747 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAPPC13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
TRAPPC13 - trafficking protein particle complex 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001093755.1 | Intron | NP_001087224.1 | ||||
NM_001093756.1 | Intron | NP_001087225.1 | ||||
NM_001243737.1 | Intron | NP_001230666.1 | ||||
NM_024941.3 | Intron | NP_079217.2 | ||||
XM_011543652.2 | Intron | XP_011541954.1 | ||||
XM_017009890.1 | Intron | XP_016865379.1 |
TRIM23 - tripartite motif containing 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001656.3 | Intron | NP_001647.1 | ||||
NM_033227.2 | Intron | NP_150230.1 | ||||
NM_033228.2 | Intron | NP_150231.1 | ||||
XM_005248497.3 | Intron | XP_005248554.1 | ||||
XM_017009444.1 | Intron | XP_016864933.1 |
Set Membership: |
HapMap Validated |