Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTTATTACCATAATGTTAGAGAT[A/G]AGTATTTTCCTTCTTACACTGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614758 MIM: 612430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCTN4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCTN4 - dynactin subunit 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135643.1 | 3088 | UTR 3 | NP_001129115.1 | |||
NM_001135644.1 | 3088 | UTR 3 | NP_001129116.1 | |||
NM_016221.3 | 3088 | UTR 3 | NP_057305.1 | |||
XM_005268457.1 | 3088 | UTR 3 | XP_005268514.1 | |||
XM_011537644.1 | 3088 | UTR 3 | XP_011535946.1 | |||
XM_011537645.1 | 3088 | UTR 3 | XP_011535947.1 | |||
XM_011537646.1 | 3088 | UTR 3 | XP_011535948.1 | |||
XM_011537647.1 | 3088 | UTR 3 | XP_011535949.1 |
RBM22 - RNA binding motif protein 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |