Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGCAGTGAACAATTCAAAGATG[G/T]TTCCCCCACAGAGTGACCTTCCCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606596 MIM: 109190 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKRP PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FKRP - fukutin related protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC1A5 - solute carrier family 1 member 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145144.1 | Intron | NP_001138616.1 | ||||
NM_001145145.1 | Intron | NP_001138617.1 | ||||
NM_005628.2 | Intron | NP_005619.1 | ||||
XM_005259167.4 | Intron | XP_005259224.1 |
Set Membership: |
HapMap |