Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616401 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SPDL1 - spindle apparatus coiled-coil protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017785.4 | 30 | Intron | NP_060255.3 | |||
XM_011534585.1 | 30 | Intron | XP_011532887.1 | |||
XM_011534586.2 | 30 | UTR 5 | XP_011532888.1 | |||
XM_011534587.2 | 30 | Intron | XP_011532889.1 | |||
XM_011534588.2 | 30 | Intron | XP_011532890.1 | |||
XM_011534589.2 | 30 | Intron | XP_011532891.1 | |||
XM_011534590.2 | 30 | Intron | XP_011532892.1 | |||
XM_017009601.1 | 30 | UTR 5 | XP_016865090.1 | |||
XM_017009602.1 | 30 | Intron | XP_016865091.1 |
Set Membership: |
HapMap Validated |