Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616401 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPDL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPDL1 - spindle apparatus coiled-coil protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017785.4 | 266 | Intron | NP_060255.3 | |||
XM_011534585.1 | 266 | Intron | XP_011532887.1 | |||
XM_011534586.2 | 266 | Intron | XP_011532888.1 | |||
XM_011534587.2 | 266 | UTR 5 | XP_011532889.1 | |||
XM_011534588.2 | 266 | UTR 5 | XP_011532890.1 | |||
XM_011534589.2 | 266 | Intron | XP_011532891.1 | |||
XM_011534590.2 | 266 | Intron | XP_011532892.1 | |||
XM_017009601.1 | 266 | Intron | XP_016865090.1 | |||
XM_017009602.1 | 266 | Intron | XP_016865091.1 |
Set Membership: |
HapMap |