Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605872 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC4M PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CLEC4M - C-type lectin domain family 4 member M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144904.1 | Intron | NP_001138376.1 | ||||
NM_001144905.1 | Intron | NP_001138377.1 | ||||
NM_001144906.1 | Intron | NP_001138378.1 | ||||
NM_001144907.1 | Intron | NP_001138379.1 | ||||
NM_001144908.1 | Intron | NP_001138380.1 | ||||
NM_001144909.1 | Intron | NP_001138381.1 | ||||
NM_001144910.1 | Intron | NP_001138382.1 | ||||
NM_001144911.1 | Intron | NP_001138383.1 | ||||
NM_014257.4 | Intron | NP_055072.3 | ||||
XM_006722611.2 | Intron | XP_006722674.1 | ||||
XM_006722612.3 | Intron | XP_006722675.1 | ||||
XM_006722613.3 | Intron | XP_006722676.1 | ||||
XM_006722614.3 | Intron | XP_006722677.1 | ||||
XM_006722615.2 | Intron | XP_006722678.1 |
Set Membership: |
HapMap Validated |