Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGCGACGGCCCAGTGGAAAGAG[A/G]ACAAACCCTTGTGGGCCTTAGGGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613478 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC106 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC106 - coiled-coil domain containing 106 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107983998 - uncharacterized LOC107983998 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527018.2 | 1414 | Missense Mutation | AAC,GAC | N,D 212 | XP_011525320.1 | |
XM_011527021.1 | 1414 | Silent Mutation | AGA,AGG | R,R 217 | XP_011525323.1 |
ZNF580 - zinc finger protein 580 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163423.1 | 1414 | Intron | NP_001156895.1 | |||
NM_016202.2 | 1414 | Intron | NP_057286.1 | |||
NM_207115.1 | 1414 | Intron | NP_996998.1 |
ZNF581 - zinc finger protein 581 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016535.3 | 1414 | Intron | NP_057619.1 | |||
XM_006723240.3 | 1414 | Intron | XP_006723303.1 | |||
XM_017026867.1 | 1414 | Intron | XP_016882356.1 |
Set Membership: |
HapMap |