Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCGCCTTGCTCGGAGGTGCCAC[A/G]GTGTTTGAAATGAAGCCTGGGGGGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613430 MIM: 610887 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAUS3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
HAUS3 - HAUS augmin like complex subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4800 - microRNA 4800 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MXD4 - MAX dimerization protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006454.2 | 2690 | UTR 3 | NP_006445.1 | |||
XM_011513388.2 | 2690 | UTR 3 | XP_011511690.1 | |||
XM_017007655.1 | 2690 | Intron | XP_016863144.1 | |||
XM_017007656.1 | 2690 | Intron | XP_016863145.1 |
POLN - DNA polymerase nu | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |