Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGCCAACCTTAACTCTTAAAACA[C/T]CTACCTTGCTGGCCAACCTGCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
70 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609764 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KDM4A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KDM4A - lysine demethylase 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014663.2 | Intron | NP_055478.2 | ||||
XM_005271354.3 | Intron | XP_005271411.1 | ||||
XM_005271355.3 | Intron | XP_005271412.1 | ||||
XM_005271356.3 | Intron | XP_005271413.1 | ||||
XM_011542467.2 | Intron | XP_011540769.2 | ||||
XM_017002912.1 | Intron | XP_016858401.1 |
Set Membership: |
HapMap |