Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTATATTTTACTGCATATACTGTA[A/G]GCTTAGATTTGGGTTGTGAGTGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616783 | ||||||||||||||||||||
Literature Links: |
PLA2G2C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLA2G2C - phospholipase A2 group IIC | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBXN10 - UBX domain protein 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152376.4 | Intron | NP_689589.1 | ||||
XM_005245742.3 | Intron | XP_005245799.1 | ||||
XM_011540699.2 | Intron | XP_011539001.1 |
UBXN10-AS1 - UBXN10 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |