Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATGCATTTATGAGCCCTGGGG[C/G]GCAGGGCGCAGGGCTGAGCAAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616793 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G2F PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G2F - phospholipase A2 group IIF | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022819.3 | Intron | NP_073730.3 | ||||
XM_011541955.1 | Intron | XP_011540257.1 | ||||
XM_011541956.1 | Intron | XP_011540258.1 |