Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604092 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TTK - TTK protein kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166691.1 | 725 | Intron | NP_001160163.1 | |||
NM_003318.4 | 725 | Intron | NP_003309.2 | |||
XM_011536099.2 | 725 | Intron | XP_011534401.1 | |||
XM_011536100.2 | 725 | Intron | XP_011534402.1 | |||
XM_017011242.1 | 725 | UTR 5 | XP_016866731.1 |
Set Membership: |
HapMap |