Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCCCAGTCACTAGTCTGGCAGA[A/G]ATCTGTTTCTTGAGAGTTTGAATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612532 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PHF13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
PHF13 - PHD finger protein 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153812.2 | Intron | NP_722519.2 | ||||
XM_011540762.1 | Intron | XP_011539064.1 |
THAP3 - THAP domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |