Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGCGAAGAGAGCGTGGTCTCGG[G/T]CTTAGCCTTGCTCTGGCCACTCGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 185641 MIM: 185640 MIM: 185620 MIM: 185630 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED22 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MED22 - mediator complex subunit 22 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133640.4 | Intron | NP_598395.1 | ||||
NM_181491.2 | Intron | NP_852468.1 |
RPL7A - ribosomal protein L7a | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000972.2 | Intron | NP_000963.1 |
SNORD24 - small nucleolar RNA, C/D box 24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD36A - small nucleolar RNA, C/D box 36A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD36B - small nucleolar RNA, C/D box 36B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD36C - small nucleolar RNA, C/D box 36C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF1 - surfeit 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SURF2 - surfeit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |