Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118960 MIM: 603824 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTA PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CLTA - clathrin light chain A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNE - glucosamine (UDP-N-acetyl)-2-epimerase/N-acetylmannosamine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128227.2 | Intron | NP_001121699.1 | ||||
NM_001190383.1 | Intron | NP_001177312.1 | ||||
NM_001190384.1 | Intron | NP_001177313.1 | ||||
NM_001190388.1 | Intron | NP_001177317.1 | ||||
NM_005476.5 | Intron | NP_005467.1 | ||||
XM_005251334.4 | Intron | XP_005251391.1 | ||||
XM_017014167.1 | Intron | XP_016869656.1 | ||||
XM_017014168.1 | Intron | XP_016869657.1 |
Set Membership: |
HapMap |