Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610534 MIM: 606252 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCPS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DCPS - decapping enzyme, scavenger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014026.4 | 969 | Silent Mutation | TTC,TTT | F,F 63 | NP_054745.1 | |
XM_011542778.2 | 969 | Silent Mutation | TTC,TTT | F,F 63 | XP_011541080.1 | |
XM_011542780.2 | 969 | Intron | XP_011541082.1 | |||
XM_017017612.1 | 969 | UTR 5 | XP_016873101.1 |
HP11014 - uncharacterized HP11014 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TIRAP - TIR domain containing adaptor protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |