Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCACTTATTTTCCTCTAAGAAC[C/G]TTTTTAGATGATTATGTCTTTCAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601300 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACVR1B - activin A receptor type 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004302.4 | Intron | NP_004293.1 | ||||
NM_020327.3 | Intron | NP_064732.3 | ||||
NM_020328.3 | Intron | NP_064733.3 | ||||
XM_011538966.2 | Intron | XP_011537268.1 | ||||
XM_011538967.2 | Intron | XP_011537269.1 | ||||
XM_017020199.1 | Intron | XP_016875688.1 | ||||
XM_017020200.1 | Intron | XP_016875689.1 | ||||
XM_017020201.1 | Intron | XP_016875690.1 |
Set Membership: |
HapMap |