Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTRNR2L4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTRNR2L4 - MT-RNR2-like 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZSCAN32 - zinc finger and SCAN domain containing 32 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284527.1 | Intron | NP_001271456.1 | ||||
NM_001284528.1 | Intron | NP_001271457.1 | ||||
NM_001284529.1 | Intron | NP_001271458.1 | ||||
NM_001324340.1 | Intron | NP_001311269.1 | ||||
NM_001324341.1 | Intron | NP_001311270.1 | ||||
NM_001324342.1 | Intron | NP_001311271.1 | ||||
NM_001324343.1 | Intron | NP_001311272.1 | ||||
NM_001324344.1 | Intron | NP_001311273.1 | ||||
NM_001324345.1 | Intron | NP_001311274.1 | ||||
NM_001324346.1 | Intron | NP_001311275.1 | ||||
NM_017810.3 | Intron | NP_060280.2 | ||||
XM_005255403.2 | Intron | XP_005255460.1 | ||||
XM_011522555.2 | Intron | XP_011520857.1 | ||||
XM_017023371.1 | Intron | XP_016878860.1 | ||||
XM_017023372.1 | Intron | XP_016878861.1 | ||||
XM_017023373.1 | Intron | XP_016878862.1 |
Set Membership: |
HapMap |