Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGAGAAAGCCAACTATTACACTT[C/T]AAATAAGCAATTCAGATAAGCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604798 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HOMER1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
|||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
HOMER1 - homer scaffolding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277077.1 | Intron | NP_001264006.1 | ||||
NM_001277078.1 | Intron | NP_001264007.1 | ||||
NM_004272.4 | Intron | NP_004263.1 | ||||
XM_017010059.1 | Intron | XP_016865548.1 |