Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGCTGTCATCATTCACAAGTCCA[C/T]TACACCTGTGTCAATGCATGTTAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616936 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHD9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHD9 - chromodomain helicase DNA binding protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308319.1 | Intron | NP_001295248.1 | ||||
NM_025134.4 | Intron | NP_079410.4 | ||||
XM_005256168.3 | Intron | XP_005256225.1 | ||||
XM_005256169.2 | Intron | XP_005256226.1 | ||||
XM_005256172.3 | Intron | XP_005256229.1 | ||||
XM_005256176.3 | Intron | XP_005256233.1 | ||||
XM_006721281.2 | Intron | XP_006721344.1 | ||||
XM_006721283.3 | Intron | XP_006721346.1 | ||||
XM_011523347.2 | Intron | XP_011521649.1 | ||||
XM_017023718.1 | Intron | XP_016879207.1 | ||||
XM_017023719.1 | Intron | XP_016879208.1 | ||||
XM_017023720.1 | Intron | XP_016879209.1 | ||||
XM_017023721.1 | Intron | XP_016879210.1 | ||||
XM_017023722.1 | Intron | XP_016879211.1 | ||||
XM_017023723.1 | Intron | XP_016879212.1 | ||||
XM_017023724.1 | Intron | XP_016879213.1 | ||||
XM_017023725.1 | Intron | XP_016879214.1 | ||||
XM_017023726.1 | Intron | XP_016879215.1 | ||||
XM_017023727.1 | Intron | XP_016879216.1 | ||||
XM_017023728.1 | Intron | XP_016879217.1 |