Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCTAATTGATAACAATGCTGAGT[C/T]TTCCTGTCCACGAACCTGGAATGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612145 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIRLET7D PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIRLET7D - microRNA let-7d | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIRLET7DHG - MIRLET7D host gene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |