Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612790 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF28 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARHGEF28 - Rho guanine nucleotide exchange factor 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080479.2 | Intron | NP_001073948.2 | ||||
NM_001177693.1 | Intron | NP_001171164.1 | ||||
NM_001244364.1 | Intron | NP_001231293.1 | ||||
XM_005248568.4 | Intron | XP_005248625.1 | ||||
XM_011543546.1 | Intron | XP_011541848.1 | ||||
XM_011543547.2 | Intron | XP_011541849.1 | ||||
XM_017009702.1 | Intron | XP_016865191.1 | ||||
XM_017009703.1 | Intron | XP_016865192.1 |