Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605312 MIM: 607518 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GDF15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GDF15 - growth differentiation factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC25 - leucine rich repeat containing 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_145256.2 | 1076 | Missense Mutation | NP_660299.2 | |||
XM_005259739.3 | 1076 | Missense Mutation | XP_005259796.1 |
MIR3189 - microRNA 3189 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |