Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCACTCTGAAAATTCAGTAATCT[A/C]GCATATGAGTATTACAGTAGAGGAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608238 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPPL2A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPPL2A - signal peptide peptidase like 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032802.3 | 6661 | Intron | NP_116191.2 | |||
XM_005254722.3 | 6661 | UTR 3 | XP_005254779.1 | |||
XM_017022680.1 | 6661 | Intron | XP_016878169.1 | |||
XM_017022681.1 | 6661 | Intron | XP_016878170.1 |
Set Membership: |
HapMap |