Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAACTAAAAGCAAGAGTCATTTCT[C/T]AGTTCTGGTCTTGCCTCCCACGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 157140 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAPT - microtubule associated protein tau | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001123066.3 | Intron | NP_001116538.2 | ||||
NM_001123067.3 | Intron | NP_001116539.1 | ||||
NM_001203251.1 | Intron | NP_001190180.1 | ||||
NM_001203252.1 | Intron | NP_001190181.1 | ||||
NM_005910.5 | Intron | NP_005901.2 | ||||
NM_016834.4 | Intron | NP_058518.1 | ||||
NM_016835.4 | Intron | NP_058519.3 | ||||
NM_016841.4 | Intron | NP_058525.1 | ||||
XM_005257362.4 | Intron | XP_005257419.1 | ||||
XM_005257364.4 | Intron | XP_005257421.1 | ||||
XM_005257365.4 | Intron | XP_005257422.1 | ||||
XM_005257366.3 | Intron | XP_005257423.1 | ||||
XM_005257367.4 | Intron | XP_005257424.1 | ||||
XM_005257368.4 | Intron | XP_005257425.1 | ||||
XM_005257369.4 | Intron | XP_005257426.1 | ||||
XM_005257370.4 | Intron | XP_005257427.1 | ||||
XM_005257371.4 | Intron | XP_005257428.1 |
MAPT-IT1 - MAPT intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |