Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCATCATCCACATATGGATTAGC[C/T]ATGTTGTGGACAGTGCAAGTACTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 176844 MIM: 602458 | ||||||||||||||||||||
Literature Links: |
PSMA5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSMA5 - proteasome subunit alpha 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199772.1 | 1974 | UTR 3 | NP_001186701.1 | |||
NM_001199773.1 | 1974 | UTR 3 | NP_001186702.1 | |||
NM_001199774.1 | 1974 | UTR 3 | NP_001186703.1 | |||
NM_002790.3 | 1974 | UTR 3 | NP_002781.2 |
SORT1 - sortilin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |