Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTCAAGCTTGGCCCAAGTGAAC[T/C]GTCTACTTACATTTGTGGTTCCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603974 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF92 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ZNF92 - zinc finger protein 92 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287532.1 | Intron | NP_001274461.1 | ||||
NM_001287533.1 | Intron | NP_001274462.1 | ||||
NM_001287534.1 | Intron | NP_001274463.1 | ||||
NM_007139.3 | Intron | NP_009070.2 | ||||
NM_152626.3 | Intron | NP_689839.1 |
Set Membership: |
HapMap Validated |