Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATCAGTTACTGTCTTTTTTTGGA[G/T]TTTTAAATTAATGATCATTTTTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182279 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRPN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNRPN - small nuclear ribonucleoprotein polypeptide N | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003097.4 | Intron | NP_003088.1 | ||||
NM_022805.3 | Intron | NP_073716.1 | ||||
NM_022806.3 | Intron | NP_073717.1 | ||||
NM_022807.3 | Intron | NP_073718.1 | ||||
NM_022808.3 | Intron | NP_073719.1 |
SNURF - SNRPN upstream reading frame | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005678.4 | Intron | NP_005669.2 | ||||
NM_022804.2 | Intron | NP_073715.1 |
Set Membership: |
HapMap |