Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605487 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPP19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARPP19 - cAMP regulated phosphoprotein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306191.1 | Intron | NP_001293120.1 | ||||
NM_001306195.1 | Intron | NP_001293124.1 | ||||
NM_001306196.1 | Intron | NP_001293125.1 | ||||
NM_006628.5 | Intron | NP_006619.1 | ||||
XM_017021868.1 | Intron | XP_016877357.1 | ||||
XM_017021869.1 | Intron | XP_016877358.1 | ||||
XM_017021870.1 | Intron | XP_016877359.1 | ||||
XM_017021871.1 | Intron | XP_016877360.1 |