Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603668 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SCAF11 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
SCAF11 - SR-related CTD associated factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004719.2 | 8391 | UTR 3 | NP_004710.2 | |||
XM_005269230.2 | 8391 | UTR 3 | XP_005269287.2 | |||
XM_011538984.2 | 8391 | UTR 3 | XP_011537286.1 | |||
XM_011538985.1 | 8391 | UTR 3 | XP_011537287.1 | |||
XM_017020217.1 | 8391 | UTR 3 | XP_016875706.1 | |||
XM_017020218.1 | 8391 | Intron | XP_016875707.1 | |||
XM_017020219.1 | 8391 | UTR 3 | XP_016875708.1 | |||
XM_017020220.1 | 8391 | UTR 3 | XP_016875709.1 | |||
XM_017020221.1 | 8391 | UTR 3 | XP_016875710.1 | |||
XM_017020222.1 | 8391 | UTR 3 | XP_016875711.1 | |||
XM_017020223.1 | 8391 | UTR 3 | XP_016875712.1 |