Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTGGGTGCCTTTCATTTGTATAT[A/G]TGGGTAAATGTACTTTAATCCTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604447 MIM: 610022 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNB5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
GNB5 - G protein subunit beta 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100129973 - uncharacterized LOC100129973 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MYO5C - myosin VC | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018728.3 | Intron | NP_061198.2 | ||||
XM_011521781.2 | Intron | XP_011520083.1 | ||||
XM_017022408.1 | Intron | XP_016877897.1 | ||||
XM_017022409.1 | Intron | XP_016877898.1 | ||||
XM_017022410.1 | Intron | XP_016877899.1 |
Set Membership: |
HapMap Validated |