Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604904 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAF1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TAF1B - TATA-box binding protein associated factor, RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318976.1 | 35 | UTR 5 | NP_001305905.1 | |||
NM_001318977.1 | 35 | UTR 5 | NP_001305906.1 | |||
NM_005680.2 | 35 | Missense Mutation | GCG,TCG | A,S 6 | NP_005671.2 | |
XM_011510410.2 | 35 | Intron | XP_011508712.1 | |||
XM_011510411.1 | 35 | UTR 5 | XP_011508713.1 |
Set Membership: |
HapMap |