Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGTTTCATGACATATGAAAAAC[A/T]TTGGGAAAATAAATTTTTCAAGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND5B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DENND5B - DENN domain containing 5B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308339.1 | 7889 | UTR 3 | NP_001295268.1 | |||
NM_144973.3 | 7889 | UTR 3 | NP_659410.3 | |||
XM_005253326.2 | 7889 | UTR 3 | XP_005253383.1 | |||
XM_005253327.3 | 7889 | UTR 3 | XP_005253384.1 | |||
XM_005253330.3 | 7889 | UTR 3 | XP_005253387.1 | |||
XM_006719040.3 | 7889 | UTR 3 | XP_006719103.1 | |||
XM_011520590.2 | 7889 | UTR 3 | XP_011518892.1 | |||
XM_017018896.1 | 7889 | UTR 3 | XP_016874385.1 | |||
XM_017018897.1 | 7889 | UTR 3 | XP_016874386.1 | |||
XM_017018898.1 | 7889 | UTR 3 | XP_016874387.1 | |||
XM_017018899.1 | 7889 | UTR 3 | XP_016874388.1 |