Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTCACATTTTTTCTCAAGATT[A/T]TTTACTGGCATCTGCACCAAAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604277 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAST PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SPAST - spastin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014946.3 | Intron | NP_055761.2 | ||||
NM_199436.1 | Intron | NP_955468.1 | ||||
XM_005264516.4 | Intron | XP_005264573.1 | ||||
XM_011533067.2 | Intron | XP_011531369.1 | ||||
XM_017004777.1 | Intron | XP_016860266.1 | ||||
XM_017004778.1 | Intron | XP_016860267.1 |
Set Membership: |
HapMap |