Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTACTTACTAGAAAAAATGAGAAG[C/T]AGATAAAGGTTATCTCTGCTGTCCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBM27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
RBM27 - RNA binding motif protein 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018989.1 | Intron | NP_061862.1 | ||||
XM_005268466.1 | Intron | XP_005268523.1 | ||||
XM_005268467.1 | Intron | XP_005268524.1 | ||||
XM_005268468.1 | Intron | XP_005268525.1 | ||||
XM_006714787.1 | Intron | XP_006714850.1 | ||||
XM_011537661.1 | Intron | XP_011535963.1 | ||||
XM_011537662.1 | Intron | XP_011535964.1 | ||||
XM_017009587.1 | Intron | XP_016865076.1 |
Set Membership: |
HapMap Validated |