Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACACTTGTTTTTGGGACTGTCCG[C/T]TCTCAGCAGCTCTCCAAGGAAAGTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613403 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM127 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM127 - transmembrane protein 127 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193304.2 | 957 | UTR 3 | NP_001180233.1 | |||
NM_017849.3 | 957 | UTR 3 | NP_060319.1 | |||
XM_017004450.1 | 957 | Missense Mutation | AAC,AGC | N,S 46 | XP_016859939.1 | |
XM_017004451.1 | 957 | UTR 3 | XP_016859940.1 | |||
XM_017004452.1 | 957 | UTR 3 | XP_016859941.1 |