Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCACTCCCCTTCCCCATTCCTG[A/G]CAGAAGAAAATGCAAAAGATCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606017 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMMECR1L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AMMECR1L - AMMECR1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199140.1 | 3366 | UTR 3 | NP_001186069.1 | |||
NM_031445.2 | 3366 | Intron | NP_113633.2 | |||
XM_005263804.4 | 3366 | UTR 3 | XP_005263861.1 | |||
XM_005263806.4 | 3366 | UTR 3 | XP_005263863.1 | |||
XM_005263807.4 | 3366 | UTR 3 | XP_005263864.1 | |||
XM_011511952.2 | 3366 | UTR 3 | XP_011510254.1 | |||
XM_011511954.2 | 3366 | UTR 3 | XP_011510256.1 | |||
XM_011511955.2 | 3366 | Intron | XP_011510257.1 | |||
XM_011511956.2 | 3366 | Intron | XP_011510258.1 | |||
XM_017005063.1 | 3366 | Intron | XP_016860552.1 |
LOC107985803 - uncharacterized LOC107985803 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017005446.1 | 3366 | Intron | XP_016860935.1 |
POLR2D - RNA polymerase II subunit D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |