Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGATTTGGCCACCAGAGGGCAG[C/G]AGCTGCAGCAGCCAGGCAGGCGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DPEP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DPEP1 - dipeptidase 1 (renal) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128141.2 | Intron | NP_001121613.1 | ||||
NM_004413.3 | Intron | NP_004404.1 | ||||
XM_005256285.4 | Intron | XP_005256342.1 | ||||
XM_005256286.3 | Intron | XP_005256343.1 | ||||
XM_011522925.2 | Intron | XP_011521227.1 | ||||
XM_017023009.1 | Intron | XP_016878498.1 |